missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 1%, Copper, 0.3% on alumina granules, reduced

Brand:  Alfa Aesar™ 044872.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11366289

  • £153.80 / 25g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
1%, Copper, 0.3% on alumina granules, Reduced
MFCD08064572

Specifications

Palladium
Granular
25g
1%, Copper, 0.3% on alumina granules, Reduced
MFCD08064572
Videos
SDS