missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 1% on activated carbon powder, standard, unreduced, nominally 50% water wet

Brand:  Alfa Aesar™ 038293.22

Code : NEW

Additional Details : Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11374387

  • £524.59 / 100g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
1% on activated carbon powder, Standard, unreduced, nominally 50% water wet
MFCD03457879

Specifications

Palladium
MFCD03457879
900 to 1100m2/g
1% on activated carbon powder, Standard, unreduced, nominally 50% water wet
100g
Videos
SDS