missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 0.12%, Ruthenium, 0.12%; on 3mm alumina tablets

Brand:  Alfa Aesar™ 042206.09

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15426445

  • £46.20 / 10g


EU Stock 1
Add to basket

Description

Description

  • Reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Specifications

Chemical Identifiers

Palladium
MFCD03613602

Specifications

Palladium
MFCD03613602
10g
7440-05-3
0.12%, Ruthenium, 0.12%; on 3mm alumin.a tablets
Videos
SDS