Thermo Scientific PageRuler™ Plus Prestained 10-250kDa Protein Ladder

Mixture of 9 blue-, orange- and green-stained proteins (10 to 250kDa) used as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

Overview
Brand: Thermo Scientific

Manufacturer Part Number: 26621

UNSPSC: 41105337

Code: PN

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11864544

Quantity Price
1 £ 15.15 / Each
Estimated Shipment date
from Supplier 04-05-2017
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Quantity 25μL
Description PageRuler Plus Prestained Protein Ladder, 10 to 250kDa
Stained Prestained
Separation Range Broad
Sufficient For 2to 5 mini gel lanes at 5 to 10uL per lane
Detection Visible
Formulation 9 proteins from 10 to 250kDa in reducing sample buffer

This prestained protein MW marker is designed for monitoring the progress of SDS-polyacrylamide gel electrophoresis, for assessing transfer efficiency onto PVDF, nylon and nitrocellulose membranes, and for estimating the approximate size of separated proteins that have been made visible with gel stains or Western blot detection reagents. A blue chromophore is bound to all proteins, except proteins of two reference bands of 70kDa and 25kDa that are colored with an orange dye and one green reference band of 10kDa. PageRuler Plus Prestained Protein Ladder is ready to use: no heating, further dilution or addition of a reducing agent is required before loading onto a gel.

Highlights:

  • Size range – nine proteins spanning 10 to 250kDa

  • Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil

  • Sharp bands – color-coded bands of similar intesity for easy visualization

  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting

  • Bright reference bands – orange at 70 and 25kDa, and green at 10kDa

  • Membrane-compatible – colored bands transfer to membranes for Western blotting

Includes:

Dye-stained proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1mM NaN3 and 33% glycerol.

Recommended for:

Monitoring protein migration during SDS-polyacrylamide gel electrophoresis; Monitoring protein transfer onto membranes after Western blotting; Sizing of proteins on SDS-polyacrylamide gels and Western blots

Alternative Products