missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Paddle Blender Bags with Full Filter

Fisherbrand Paddle Blender Bags for use with Paddle Blenders.

Brand:  Fisherbrand™

Additional Details : Weight : 2.29900kg

Product Code. 17379912

  • £545.00 / Pack of 500
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

  • Sterile, 400 mL bags
  • High quality, virgin LDPE
  • Resistant to rips and tears
  • Fit most paddle blenders
  • Available with or without filter
Specifications

Specifications

400 mL
Food and other samples
Paddle Blenders
300 x 190 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers