Learn More
Molecular Probes™ Pacific Orange™ Succinimidyl Ester, Triethylammonium Salt
Pacific Orange™ Succinimidyl Ester, Triethylammonium Salt
Brand: Molecular Probes™ P30253
Code : 50
Additional Details : Weight : 0.00525kg
Description
Pacific Orange™ succinimidyl ester is a violet-excitable (405 nm) dye with emission maximum at ∼551 nm. This allows it to be used with other violet-excitable fluorophores such as Pacific Blue™ dye for multiparameter analysis using a single excitation source. As a class of compounds, succinimidyl ester forms provide the most efficient and easy-to-use reaction chemistry to selectively link a dye to accessible primary amine groups on proteins, modified nucleic acids, or other biomolecules. Succinimidyl esters are excellent reagents for amine modification because the covalent bonds they form are as stable as the peptide bonds used to link amino acids in proteins. Pacific Orange™ succinimidyl ester is supplied as 1 mg of dry powder and should be stored at ≤-20°C, desiccated, and protected from light.
Specifications
1 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Pacific Orange™ | |
Pacific Orange™ | |
405 |
Succinimidyl Ester | |
Amine | |
Active Ester, Succinimidyl Ester | |
Pacific Dyes | |
551 |
For Research Use Only. Not for use in diagnostic procedures.