missing translation for 'onlineSavingsMsg'
Learn More

P7C3, Tocris Bioscience™

NAMPT activator; also proneurogenic and neuroprotective

Brand:  Tocris (Bio-Techne) 407610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13454084

  • £183.75 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NAMPT activator. Proneurogenic and neuroprotective. Protects newborn neurons in the dentate gyrus by mitigating cell death. Also enhances learning and memory in aged rats. Orally available and brain penetrant. Also revents doxorubicin-mediated toxicity in osteosarcoma cells and restores intracellular NAD levels.
Specifications

Chemical Identifiers

301353-96-8
474.196
1-3,6-dibromo-carbazol-9-yl-3-phenylamino-propan-2-ol, 1-3,6-dibromo-9h-carbazol-9-yl-3-phenylamino propan-2-ol, 1-anilino-3-3,6-dibromocarbazol-9-yl propan-2-ol, 1-3,6-dibromocarbazol-9-yl-3-phenylamino propan-2-ol, 3,6-dibromo-alpha-phenylamino methyl-9h-carbazole-9-ethanol, acmc-20drae, p7c hplc, 3,6-dibromo-?-phenylamino methyl-9h-carbazole-9-ethanol, 3,6-dibromo-a-phenylamino methyl-9h-carbazole-9-ethanol
1-anilino-3-(3,6-dibromocarbazol-9-yl)propan-2-ol
C21H18Br2N2O
FZHHRERIIVOATI-UHFFFAOYSA-N
2836187
C1=CC=C(C=C1)NCC(CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)O

Specifications

10mg
301353-96-8
1-3,6-dibromo-carbazol-9-yl-3-phenylamino-propan-2-ol, 1-3,6-dibromo-9h-carbazol-9-yl-3-phenylamino propan-2-ol, 1-anilino-3-3,6-dibromocarbazol-9-yl propan-2-ol, 1-3,6-dibromocarbazol-9-yl-3-phenylamino propan-2-ol, 3,6-dibromo-alpha-phenylamino methyl-9h-carbazole-9-ethanol, acmc-20drae, p7c hplc, 3,6-dibromo-?-phenylamino methyl-9h-carbazole-9-ethanol, 3,6-dibromo-a-phenylamino methyl-9h-carbazole-9-ethanol
C1=CC=C(C=C1)NCC(CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)O
474.196
474.19
P7C3
C21H18Br2N2O
FZHHRERIIVOATI-UHFFFAOYSA-N
1-anilino-3-(3,6-dibromocarbazol-9-yl)propan-2-ol
2836187
>99%
Videos
Documents