missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Ab-6 Mouse Monoclonal Antibody, Epredia™
Achieve accurate, reproducible results with the Epredia™ p53 Ab-6, Mouse Monoclonal Antibody.
£117.30 - £444.72
Specifications
Antigen | p53 Ab-6 |
---|---|
Clone | DO-1 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12670377
|
Epredia
MS-187-P1ABX |
Concentrated | 100 μL |
£347.00
100µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690377
|
Epredia
MS-187-P1 |
Concentrated | 500 μL |
£310.00
500µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12600387
|
Epredia
MS-187-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12680377
|
Epredia
MS-187-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12660377
|
Epredia
MS-187-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.Host Species: Mouse
Clone: DO-1
Isotype: IgG2a
Species Reactivity: Human, Monkey and Cow. Reacts weakly with Mouse and Rat. Others not known.
Epitope: aa 11-25
Immunogen: Recombinant human wild-type p53 protein expressed in E. coli
Molecular Weight: 53kDa
Positive Control: SKBR-3 cells or MDA-231 cells, Colon CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
- Gel Supershift Assay
Note:
USA: RUO; Int'l: RUO
Specifications
p53 Ab-6 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Cell Cycle | |
Recombinant human wild-type p53 protein expressed in E. coli | |
Primary |
DO-1 | |
Monoclonal | |
Concentrated | |
RUO | |
Bovine, Human, Non-human Primate | |
IgG2a |