missing translation for 'onlineSavingsMsg'
Learn More

p53 Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Achieve accurate, reproducible results with the Epredia™ p53 Ab-2, Mouse Monoclonal Antibody.

£104.04 - £416.16

Specifications

Antigen p53 Ab-2
Clone PAb 1801
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Concentration Quantity Price Quantity & Availability  
Product Code Brand Concentration Quantity Price Quantity & Availability  
12688756
View Documents Promotion Details
Epredia
MS-105-P1
200μg/mL 500 μL
£313.00
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12678756
View Documents Promotion Details
Epredia
MS-105-P0
200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12668756
View Documents Promotion Details
Epredia
MS-105-P
200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12698756
View Documents Promotion Details
Epredia
MS-105-P1ABX
1mg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12618766
View Documents Promotion Details
Epredia
MS-105-PABX
1mg/mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 1801

Isotype: IgG1

Species Reactivity: Human specific.

Epitope: aa 32-79

Immunogen: Recombinant human p53 protein

Molecular Weight: 53kDa

Positive Control: SKBR-3 cells, MDA-231 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

p53 Ab-2
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Cell Cycle
Recombinant Human p53 protein
Primary
PAb 1801
Monoclonal
Concentrated
RUO
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications