Learn More
P4HA2 Monoclonal Antibody (CL0351), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524599
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LKEYILVEEA KLSKIKSWAN KMEALTSKSA ADAEGYLAHP VNAYKLVKRL NTDWPALEDL VLQDSAAGFI ANLSVQRQFF PTDEDEIGAA KALMRLQDTY RLDPGTISRG ELPGTKYQAM LSVDDCFGM Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%. Binds to an epitope located within the peptide sequence NTDWPALEDLVLQDS as determined by overlapping synthetic peptides.
P4HA2 Monoclonal antibody specifically detects P4HA2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
P4HA2 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15460 | |
P4HA2 | |
Recombinant protein corresponding to Human P4HA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL0351 | |
Unconjugated | |
P4HA2 | |
4-PH alpha 2; 4-PH alpha-2; 4PHalpha2; AA407196; C76437; collagen prolyl 4-hydroxylase alpha(II); C-P4Halpha(II); hypothetical protein LOC507327; P4HA2; P4hl; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide 2; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; prolyl 4-hydroxylase A; Prolyl 4-hydroxylase alpha IIa subunit; prolyl 4-hydroxylase subunit alpha 2; prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II; UNQ290/PRO330; zgc:91948 | |
Mouse | |
Protein A | |
RUO | |
8974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only