missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ P450 Demethylation Fluorescent Activity Kit

Assay P450 Demethylation Activity in liver microsomes or cerosomes such as Cyp P450 3A4, 2B4 and 2D6.

Brand:  Invitrogen™ EIAP450DMT

Additional Details : Weight : 0.33820kg

Product Code. 15890903

  • £342.00 / Pack of 2


Estimated Shipment date
from Supplier 22-06-2020
Add to basket

Includes: 96 Well Plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Description

Description

  • Assay Type: Enzyme Activity kit
  • Sample Types: liver microsomes or cerosomes such as Cyp P450 3A4, 2B4 and 2D6
  • Standard Curve Range: 1.6 pmoles/100uL–20 pmoles/100uL
Specifications

Specifications

Approved for shipment on Wet or Dry Ice
Microplate Reader
96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Store at –20°C.
P450 Demethylation
Activity Kit
Videos
Documents