missing translation for 'onlineSavingsMsg'
Learn More

P1075, Tocris Bioscience™

Potent Kir6 (KATP) channel opener

Brand:  Tocris (Bio-Techne) 135550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459213

  • £665.57 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Potent Kir6 (KATP) channel opener (EC50 for relaxation of rat aorta = 7.5 nM). Binds to SUR2A and SUR2B with high affinity (Kd values are 17 and 3 nM respectively).
Specifications

Chemical Identifiers

60559-98-0
231.303
2-cyano-1-tert-pentyl-3-3-pyridyl guanidine, chembl11458, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridylguanidine, guanidine, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl guanidine, guanidine, 2-cyano-1-tert-pentyl-3-3-pyridyl, guanidine, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl-cas, tocris-1355, d00lwf, n'-cyano-n-tert-pentyl-n'-3-pyridylguanidine
1-cyano-2-(2-methylbutan-2-yl)-3-pyridin-3-ylguanidine
C12H17N5
HKZNADVVGXKQDL-UHFFFAOYSA-N
43345
CCC(C)(C)N=C(NC#N)NC1=CN=CC=C1

Specifications

P1075
60559-98-0
2-cyano-1-tert-pentyl-3-3-pyridyl guanidine, chembl11458, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridylguanidine, guanidine, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl guanidine, guanidine, 2-cyano-1-tert-pentyl-3-3-pyridyl, guanidine, n-cyano-n'-1,1-dimethylpropyl-n-3-pyridinyl-cas, tocris-1355, d00lwf, n'-cyano-n-tert-pentyl-n'-3-pyridylguanidine
CCC(C)(C)N=C(NC#N)NC1=CN=CC=C1
231.303
231.3
50mg
C12H17N5
HKZNADVVGXKQDL-UHFFFAOYSA-N
1-cyano-2-(2-methylbutan-2-yl)-3-pyridin-3-ylguanidine
43345
>99%
Videos