missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-201804
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
622-47-9
C9H10O2
150.18
MFCD00004353
GXXXUZIRGXYDFP-UHFFFAOYSA-N
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
248474
2-(4-methylphenyl)acetic acid
CC1=CC=C(C=C1)CC(=O)O
This item is not returnable. View return policy
622-47-9
C9H10O2
150.18
MFCD00004353
GXXXUZIRGXYDFP-UHFFFAOYSA-N
p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
248474
2-(4-methylphenyl)acetic acid
CC1=CC=C(C=C1)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90°C to 93°C
Color White
Boiling Point 265°C to 267°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4CH2CO2H
Quantity 5 g
Beilstein 09, 530
Formula Weight 150.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material p-Tolylacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Tolylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'