missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7081495
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
622-59-3
C8H7NS
149.211
MFCD00004813
ABQKHKWXTUVKGF-UHFFFAOYSA-N
p-tolyl isothiocyanate, 4-methylphenyl isothiocyanate, 4-methylphenylisothiocyanate, p-tolylisothiocyanate, 4-tolylisothiocyanate, benzene, 1-isothiocyanato-4-methyl, 4-tolyl isothiocyanate, isothiocyanic acid, p-tolyl ester, 1-isothiocyanato-4-methyl-benzene, isothiocyanic acid, 4-tolyl ester
12149
1-isothiocyanato-4-methylbenzene
CC1=CC=C(C=C1)N=C=S
This item is not returnable. View return policy
622-59-3
C8H7NS
149.211
MFCD00004813
ABQKHKWXTUVKGF-UHFFFAOYSA-N
p-tolyl isothiocyanate, 4-methylphenyl isothiocyanate, 4-methylphenylisothiocyanate, p-tolylisothiocyanate, 4-tolylisothiocyanate, benzene, 1-isothiocyanato-4-methyl, 4-tolyl isothiocyanate, isothiocyanic acid, p-tolyl ester, 1-isothiocyanato-4-methyl-benzene, isothiocyanic acid, 4-tolyl ester
12149
1-isothiocyanato-4-methylbenzene
CC1=CC=C(C=C1)N=C=S

p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

Solubility
Decompose in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY
Melting Point 23°C to 26°C
Boiling Point 236°C to 238°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6345
Quantity 5 g
UN Number UN2923
Beilstein 386032
Sensitivity Moisture sensitive
Solubility Information Decompose in water.
Formula Weight 149.22
Percent Purity 97%
Chemical Name or Material p-Tolyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'