missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolyl chlorothionoformate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2'-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2′-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.
Solubility
Miscible with common organic solvents.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
p-Tolyl chlorothionoformate is used in dehydration reactions by cyclic elimination of thiocarbonate esters. It acts as a catalyst in the synthesis of alfa-L-2′-deoxythreofuranosyl nucleoside analogs and alkenes from hindered alcohols.
Solubility
Miscible with common organic solvents.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 1.21 |
Boiling Point | 70°C to 72°C (1 mmHg) |
Flash Point | 96°C (204°F) |
Refractive Index | 1.574 |
Quantity | 10 g |
UN Number | UN3265 |
Beilstein | 1448986 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with common organic solvents. |
Formula Weight | 186.66 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'