missing translation for 'onlineSavingsMsg'
Learn More

p-Tolunitrile, 98+%, Thermo Scientific Chemicals

Catalog Number p-201354
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
104-85-8
C8H7N
117.15
MFCD00001827
VCZNNAKNUVJVGX-UHFFFAOYSA-N
p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
7724
4-methylbenzonitrile
CC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
104-85-8
C8H7N
117.15
MFCD00001827
VCZNNAKNUVJVGX-UHFFFAOYSA-N
p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
7724
4-methylbenzonitrile
CC1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 26.0°C to 28.0°C
Density 0.9800g/mL
Boiling Point 103.0°C to 106.0°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CN
Refractive Index 1.5285 to 1.5305
Quantity 5 g
Beilstein 09, 489
Merck Index 15, 9698
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 117.15
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material p-Tolunitrile, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'