missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-7050329
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
6192-52-5
C7H10O4S
190.213
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O
This item is not returnable. View return policy
6192-52-5
C7H10O4S
190.213
MFCD00142137
KJIFKLIQANRMOU-UHFFFAOYSA-N
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
521998
4-methylbenzenesulfonic acid;hydrate
CC1=CC=C(C=C1)S(=O)(=O)O.O

p-Toluenesulfonic acid monohydrate is used as a catalyst in the synthesis of resveratrol, in oxane derivatives as an antimalarial agent, as substituted piperidine and unsymmetrical benzyl. It acts as a catalyst in the preparation of 1,3,5-trisubstituted pyrazoles derivatives, selenated ketene dithioacetals, triazoloquinazolinone and benzimidazoquinazolinone derivatives. It also serves as an intermediate in the esterification and in reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 103°C to 106°(anhy)
Boiling Point 140°C (20 mm)
Assay Percent Range 97%
UN Number UN2585
Beilstein 3568023
Merck Index 14,9533
Formula Weight 190.22 (172.20 Anhydrous)
Percent Purity 97%
Density 1.24 g/mL
Quantity 25 g
Flash Point 150°C (302°F)
Physical Form Crystalline
Sensitivity Hygroscopic
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'