missing translation for 'onlineSavingsMsg'
Learn More

p-Tolualdehyde, 99+%, Thermo Scientific Chemicals

Catalog Number p-201151
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
104-87-0
C8H8O
120.15
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
104-87-0
C8H8O
120.15
MFCD00006954
FXLOVSHXALFLKQ-UHFFFAOYSA-N
p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde
7725
CHEBI:28617
4-methylbenzaldehyde
CC1=CC=C(C=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -6.0°C
Color Colorless to Yellow
Density 1.0150g/mL
Boiling Point 204.0°C to 205.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CHO
Refractive Index 1.5420 to 1.5480
Quantity 5 g
Beilstein 07,297
Specific Gravity 1.015
Solubility Information Solubility in water: 0.25g/l (25°C). Other solubilities: soluble in alcohol and ether
Formula Weight 120.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Tolualdehyde, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'