missing translation for 'onlineSavingsMsg'
Learn More

p-Thiocresol, 98%, Thermo Scientific Chemicals

Catalog Number p-7027908
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
106-45-6
C7H8S
124.201
MFCD00004851
WLHCBQAPPJAULW-UHFFFAOYSA-N
p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
7811
4-methylbenzenethiol
CC1=CC=C(C=C1)S
This item is not returnable. View return policy
106-45-6
C7H8S
124.201
MFCD00004851
WLHCBQAPPJAULW-UHFFFAOYSA-N
p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
7811
4-methylbenzenethiol
CC1=CC=C(C=C1)S

p-Thiocresol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Thiocresol is used as a pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Melting Point 40°C to 44°C
Boiling Point 195°C
Flash Point 68°C (154°F)
Odor Stench
Quantity 25 g
UN Number UN2811
Beilstein 605761
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material p-Thiocresol
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'