missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Phenylenediamine, 99+%, Thermo Scientific Chemicals
21.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
20.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
20.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 139.0°C to 141.0°C |
Color | Brown to Purple or Gray-White |
Flash Point | 156°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Linear Formula | C6H4(NH2)2 |
Quantity | 5 g |
Beilstein | 13, 61 |
Merck Index | 15, 7397 |
Show More |
Safety and Handling
Product Identifier
- p-Phenylenediamine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H410-Very toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'