missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents

Catalog Number p-27366
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Amber Glass
This item is not returnable. View return policy
4264-83-9
C6H4NNa2O6P
263.05
MFCD00007319
VIYFPAMJCJLZKD-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
This item is not returnable. View return policy
4264-83-9
C6H4NNa2O6P
263.05
MFCD00007319
VIYFPAMJCJLZKD-UHFFFAOYSA-L
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
p-Nitrophenyl Phosphate is suitable for use as a substrate for alkaline and acid phosphatase.
TRUSTED_SUSTAINABILITY
Melting Point 300°C
Color White
Assay Percent Range ≥98.0%
Packaging Amber Glass
Quantity 1 g
Formula Weight 371
Physical Form Powder or Solid
Chemical Name or Material p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'