missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dithiane-2,5-diol, 96%, Thermo Scientific Chemicals
71.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 130.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Plastic bottle |
Quantity | 250 g |
Solubility Information | Solubility in water: 3.9g/L (20°C). |
Formula Weight | 152.23 |
Percent Purity | 96% |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'