missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dioxane, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
489.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Specifications
Formula Weight | 88.11 |
Grade | BAKER ANALYZED™ ACS Reagent |
Quantity | 1 L |
Flash Point | 12.00°C |
Packaging | Aluminum Bottle |
Specific Gravity | 1L = 1.03kg |
Chemical Name or Material | p-Dioxane |
Safety and Handling
Signal Word
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'