missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 99+%, pure, Thermo Scientific Chemicals

Catalog Number 10701904
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
This item is not returnable. View return policy
106-44-5
C7H8O
108.14
MFCD00002376
IWDCLRJOBJJRNH-UHFFFAOYSA-N
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
2879
CHEBI:17847
4-methylphenol
CC1=CC=C(C=C1)O
This item is not returnable. View return policy
106-44-5
C7H8O
108.14
MFCD00002376
IWDCLRJOBJJRNH-UHFFFAOYSA-N
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
2879
CHEBI:17847
4-methylphenol
CC1=CC=C(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 32.0°C to 34.0°C
Color Colorless to Yellow
Density 1.0340g/mL
Boiling Point 202.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4OH
Quantity 2.5 kg
Beilstein 06,389
Merck Index 15,2564
Specific Gravity 1.034
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
Viscosity 8 mPa.s (35°C)
Formula Weight 108.14
Percent Purity 99+%
Grade Pure
Physical Form Crystalline Solid or Liquid
Chemical Name or Material p-Cresol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Cresol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'