Learn More
P-cadherin Monoclonal Antibody (12H6), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9873-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal antibody 12H6 recognizes human P-cadherin (placental-cadherin), a calcium dependent cell-cell adhesion protein. This 118 kDa transmembrane protein was originally named due to its expression in mouse placental tissue. Unfortunately, it is not expressed in human placental tissue. The function of the molecule will depend on the associations with the specific member of the cytoplasmic catenin family (alpha, beta and gamma). The specific interaction defines the strength and signaling of the cell-cell interaction. Expression is less broad than E-cadherin; P-cadherin is restricted to the basal proliferative cell layer of stratified epithelia. During development expression is found in the proliferating tissues such as hair follicle keratinocytes and the growth regions in ductal mammary tissue. There is evidence that P-cadherin can be secreted from epithelial cells during the late stages of pregcy. P-cadherin expression is associated with poor prognosis for breast cancer while reduced expression in melanomas and squamous cell carcinomas may also result in poor prognosis. Applications Reported: This 12H6 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections (IHC-P), and immunocytochemical staining (ICC). Applications Tested: This 12H6 antibody has been tested immunohistochemistry on formalin-fixed paraffin embedded (FFPE) human skin with low pH antigen retrieval.
P-cadherin, also known as Cadherin-3, is a Type 1 cadherin protein that belongs to the cadherin superfamily. Type 1 cadherins are single-pass transmembrane proteins that have 5 extracellular cadherin repeats and an intracellular domain that binds p120-catenin and beta-catenin. Cadherins are calcium-dependent cell-cell adhesion glycoproteins responsible for a range of processes including development, wound healing, cell-cell signaling, cell growth and differentiation. P-cadherin is expressed in human fetal structures, and in adult tissues such as the basal layer of the epidermis, breast, prostate, ovary, cervix, hair follicle, and corneal endothelium. P-cadherin expression has also been reported on embryonic stem cells, and stem cells of the normal mammary gland, and hair follicle. Overexpression of P-cadherin has been associated with poor prognosis in breast, prostate, ovary, colon, and stomach carcinomas. Mutations in CDH3 are associated with congential hypotrichosis with juvenile macular dystrophy.Specifications
P-cadherin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P22223 | |
Cdh3 | |
Affinity chromatography | |
RUO | |
1001 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
12H6 | |
Unconjugated | |
Cdh3 | |
AI385538; CADH3; cadherin 15, M-cadherin (myotubule); cadherin 3; cadherin 3, type 1, P-cadherin (placental); cadherin-3; Cadp; calcium-dependent adhesion protein, placental; CDH3; CDHP; HJMD; PCAD; P-cadherin; Placental cadherin; Placental-cadherin; RPE-specific cadherin | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.