missing translation for 'onlineSavingsMsg'
Learn More

p-Anisoyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-61053
Click to view available options
Quantity:
5 g
100 g
500 g
This item is not returnable. View return policy
100-07-2
C8H7ClO2
170.59
MFCD00000687
MXMOTZIXVICDSD-UHFFFAOYSA-N
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
7477
4-methoxybenzoyl chloride
COC1=CC=C(C=C1)C(Cl)=O
This item is not returnable. View return policy
100-07-2
C8H7ClO2
170.59
MFCD00000687
MXMOTZIXVICDSD-UHFFFAOYSA-N
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
7477
4-methoxybenzoyl chloride
COC1=CC=C(C=C1)C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 22°C
Color Colorless to Brown
Density 1.2600g/mL
Boiling Point 145°C (14.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCl
Refractive Index 1.5800 to 1.582
Quantity 5 g
Beilstein 10, 163
Merck Index 15, 665
Specific Gravity 1.26
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in diethyl ether,acetone and benzene
Formula Weight 170.6
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material p-Anisoyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Anisoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'