missing translation for 'onlineSavingsMsg'
Learn More

p-Anisic Acid 98%, ACROS Organics™

Brand:  Acros Organics 152372500

Code : 29

Additional Details : CAS Number : 100-09-4 Weight : 0.30000kg

Quantity 250g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10686542

  • £20.80 / 250g


EU Stock 5
Add to basket

Specifications

Chemical Identifiers

p-Anisic acid
100-09-4
98%
CH3OC6H4CO2H
MFCD00002542
98%
98%
C8H8O3
152.15
4-Methoxybenzoic acid

Specifications

p-Anisic acid
100-09-4
Powder
98%
CH3OC6H4CO2H
152.15
1.5710 to 1.5760
White to slightly gray-beige
181.0° to 186.0°C
185°C
98%
Authentic
98%
C8H8O3
Plastic bottle
MFCD00002542
4-Methoxybenzoic acid
275.0°C
250g
Videos
SDS