Learn More
Ozagrel hydrochloride, Tocris Bioscience™
Selective thromboxane A2 synthetase inhibitor
Brand:  Tocris (Bio-Techne) 151010
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13429583
  • £89.67 / 10mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Potent and selective inhibitor of thromboxane (TXA2) synthetase (IC50 = 4 nM). Does not inhibit prostacyclin (PGI2) synthetase, cyclooxygenase or PGE2 isomerase (IC50 > 1 mM). Inhibits platelet aggregation in vitro and arachidonate-induced arterial contraction in vivo.
Specifications
Chemical Identifiers
74003-18-2
264.709
e-3-4-1h-imidazol-1-yl methyl phenyl acrylic acid hydrochloride, 3-4-imidazol-1-ylmethyl phenyl prop-2-enoic acid hydrochloride
CHEBI:31954
C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl
C13H13ClN2O2
CWKFWBJJNNPGAM-IPZCTEOASA-N
6438130
(E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride
Specifications
10mg
74003-18-2
e-3-4-1h-imidazol-1-yl methyl phenyl acrylic acid hydrochloride, 3-4-imidazol-1-ylmethyl phenyl prop-2-enoic acid hydrochloride
C1=CC(=CC=C1CN2C=CN=C2)C=CC(=O)O.Cl
264.709
CHEBI:31954
>98%
Ozagrel hydrochloride
C13H13ClN2O2
CWKFWBJJNNPGAM-IPZCTEOASA-N
(E)-3-[4-(imidazol-1-ylmethyl)phenyl]prop-2-enoic acid;hydrochloride
6438130
264.71