missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxonic acid potassium salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.
Solubility
Soluble in hot water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.
Solubility
Soluble in hot water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 300°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Merck Index | 14,6947 |
Solubility Information | Soluble in hot water. |
IUPAC Name | potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate |
Formula Weight | 195.18 |
Chemical Name or Material | Oxonic acid potassium salt, 98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'