Learn More
Thermo Scientific™ Oxoid™ STAA Selective Supplement
The Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
Brand: Thermo Scientific™ SR0151E
Code : 65
Additional Details : Weight : 0.17500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
- Add to STAA Agar Base, Part No. CM0881B
- Each vial supplements 500mL of medium
Specifications
STAA Selective Supplement | |
Selective Supplement | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |