Learn More
Thermo Scientific™ Oxoid™ Skim Milk Powder
Oxoid Skim Milk Powder is a thermophile-free, spray dried powder for inclusion in bacteriological culture media.
Brand:  Thermo Scientific™ LP0031B
Code : 64
Additional Details : Weight : 0.50000kg
Product Code. 10651135
  • £22.90 / 500g
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Industrial Reference: NMKL.
Powder
500g
Skim Milk Powder
Each
Laboratory Preparations