missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Listeria Enrichment Broth

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.

Brand:  Thermo Scientific™ CM0862B

Code : 64

Additional Details : Weight : 0.01000kg

Product Code. 10127662

  • £107.00 / 500g
In Stock
to see stock.
Add to basket

Description

Description

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.

Also available Listeria Selective Supplement, Part No. SR0140E or SR0141E and Modified Listeria Selective Enrichment Supplement, Part No. SR0206E or SR0206B.
Specifications

Specifications

Industrial Reference: FDA, IDF.
Powder
500 g
Listeria Enrichment Broth
Dehydrated Culture Media
For 13.9L medium
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only