Learn More
Thermo Scientific™ Oxoid™ Campylobacter Agar Base
Oxoid Campylobacter Agar Base isolates Campylobacter jejuni and Campylobacter coli.
Brand: Thermo Scientific™ CM0689B
Code : 64
Additional Details : Weight : 0.50000kg
Description
Oxoid Campylobacter Agar Base is used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian and environmental specimens.
Also available Campylobacter Selective Supplement (Blaser-Wang), Part No. SR0098E NMKL; Bolton Broth Selective Supplement, Part No. SR0183E; Modified Bolton Broth Selective Supplement, Part No. SR0208E; Modified Preston Campylobacter Selective Supplement, Part No. SR0204E; and Campylobacter Selective Supplement Preston, Part No. SR0117E.Specifications
Campylobacter Agar Base | |
Agar | |
For 13.5L medium |
Powder | |
500 g |
For Laboratory Use Only