Haldenwanger™ Oxide Ceramic Conical Tall Form Alsint Crucibles


Overview
Brand: Haldenwanger™

Manufacturer Part Number: C/4

UNSPSC: 41120000

Code: 02

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10500062

Quantity Price
1 £ 52.85 / Each
Estimated Shipment
11-04-2017
Add to basket


Description and Specification

Specification

Product Type Alsint Crucible
Material Oxide Ceramic
Capacity (Metric) 80mL
Diameter (Metric) 50mm
Shape Conical