Learn More
Oxetan-3-ol, 95%, ACROS Organics™
Brand: Acros Organics 435840050
Code : 29
Additional Details : CAS Number : 7748-36-9 Weight : 0.05500kg
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
Oxetan-3-ol | |
95% | |
74.08 |
≥94.0% | |
C3H6O2 | |
3-Hydroxyoxetane |
Specifications
Oxetan-3-ol | |
1.1250g/mL | |
Liquid | |
95% | |
74.08 | |
3-Hydroxyoxetane | |
5g |
7748-36-9 | |
Authentic | |
≥94.0% | |
C3H6O2 | |
Glass bottle | |
Clear colorless |