missing translation for 'onlineSavingsMsg'
Learn More

Oxamide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081549
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
471-46-5
C2H4N2O2
88.066
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N
This item is not returnable. View return policy
471-46-5
C2H4N2O2
88.066
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N

Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point ∼350°C (decomposition)
Density 1.667
Odor Odorless
Quantity 100 g
Beilstein 1743262
Merck Index 14,6918
Solubility Information Soluble in ethanol,slightly soluble in water and insoluble in diethyl ether.
Formula Weight 88.07
Percent Purity ≥98%
Chemical Name or Material Oxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'