missing translation for 'onlineSavingsMsg'
Learn More

Oxamic hydrazide, 99%, Thermo Scientific Chemicals

Catalog Number p-177749
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
515-96-8
C2H5N3O2
103.08
MFCD00008004
MOKRDWKSHLLYKM-UHFFFAOYSA-N
oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
68200
2-hydrazinyl-2-oxoacetamide
C(=O)(C(=O)NN)N
This item is not returnable. View return policy
515-96-8
C2H5N3O2
103.08
MFCD00008004
MOKRDWKSHLLYKM-UHFFFAOYSA-N
oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
68200
2-hydrazinyl-2-oxoacetamide
C(=O)(C(=O)NN)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 216.0°C to 218.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCOCONHNH2
Quantity 25 g
Beilstein 02, 559
Merck Index 15, 8584
Formula Weight 103.08
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Oxamic hydrazide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'