missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10497900
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
79-37-8, 75-44-5, 76-02-8
C2Cl2O2
126.93
MFCD00000704
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
65578
oxalyl dichloride
C(=O)(C(=O)Cl)Cl
79-37-8, 75-44-5, 76-02-8
C2Cl2O2
126.93
MFCD00000704
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
65578
oxalyl dichloride
C(=O)(C(=O)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point -12.0°C
Color Colorless to Yellow
Density 1.4780g/mL
Boiling Point 63.0°C to 64.0°C (763.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCOCl
Refractive Index 1.4285 to 1.4305
Quantity 100 g
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Merck Index 15, 7013
Specific Gravity 1.478
Solubility Information Solubility in water: reacts. Other solubilities: soluble in the main common organic solvents :, methylene chloride; dichloroethane; chlorobenzene;, ethylacetate; hexane; acetone; toluene; ether;, benzene; chloroform and ether
Formula Weight 126.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Oxalyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Oxalic acid dihydrate, Oxalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'