missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Solution 0.05M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Catalog Number p-7073156
Click to view available options
Quantity:
1 Ampule
6 Ampules
This item is not returnable. View return policy
2573
This item is not returnable. View return policy
2573
pH 1
Melting Point >160°C
Quantity 1 Ampule
Packaging Ampoule
Physical Form Liquid
compliance-icons
Product Identifier
  • Oxalic acid 0.05M solution (0.5M Concentrate)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'