missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, Certified AR for Analysis, Fisher Chemical™
102.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
102.87 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
102.87 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Packaging | Plastic powder jar |
Quantity | 500 g |
Color | White |
Formula Weight | 126.04g/mol |
Vapor Pressure | 21.5mbar 50 |
pH | 1.3 |
Melting Point | 98°C |
Physical Form | Powder/Solid |
Safety and Handling
Product Identifier
- Oxalic acid dihydrate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'