missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 10% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number p-4408521
Click to view available options
Quantity:
1 L
4 L
144-62-7
C2H2O4
90.034
MFCD00002573
MUBZPKHOEPUJKR-UHFFFAOYSA-N
971
CHEBI:16995
oxalic acid
C(=O)(C(=O)O)O
144-62-7
C2H2O4
90.034
MFCD00002573
MUBZPKHOEPUJKR-UHFFFAOYSA-N
971
CHEBI:16995
oxalic acid
C(=O)(C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Color Colorless
Formula Weight 90.04
Odor Odorless
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Oxalic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'