missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40/TNFRSF4 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | OX40/TNFRSF4 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse OX40/TNFRSF4 Val20-Pro211 Accession # P47741 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
OX40/TNFRSF4 Polyclonal specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Western Blot, Agonist Activity.Specifications
OX40/TNFRSF4 | |
Goat | |
Polyclonal | |
IgG | |
ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Agonist Activity | |
RUO |
Mouse myeloma cell line NS0-derived recombinant mouse OX40/TNFRSF4 Val20-Pro211 Accession # P47741 | |
Primary | |
Mouse | |
Unconjugated | |
7293 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only