Learn More
Genlab™ Oven General Purpose Horizontal with Digital Temperature Control And Fan
Brand:  Genlab™ OV/50/DIG/F
Code : 50
Additional Details : Weight : 31.00000kg
Product Code. 12908738
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
230V 50/60Hz 730mm x 470mm x 570mm (w x d x h), 50L +40°C to 250°C 2 shelves, Genlab