missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVCA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567068
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: EAVVYLGDGRF HLESVMIANP NVPAYRYDPY SKVLSREHYD HQRMQAARQE AIATARSAK Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
OVCA1 Polyclonal antibody specifically detects OVCA1 in Human samples. It is validated for Immunocytochemistry.Specifications
OVCA1 | |
Polyclonal | |
Unconjugated | |
DPH1 | |
2-(3-amino-3-carboxypropyl)histidine synthase subunit 1; 2310011M22Rik; 4930488F09Rik; AW551873; candidate tumor suppressor in ovarian cancer 1; DEDSSH; diphthamide biosynthesis 1; diphthamide biosynthesis protein 1; diphthamide biosynthesis protein 1; 2-(3-amino-3-carboxypropyl)histidine synthase subunit 1; diphthamide biosynthesis protein 2 homolog-like 1; diphtheria toxin resistance protein 1; diptheria toxin resistance protein required for diphthamide biosynthesis (Saccharomyces)-like 1; diptheria toxin resistance protein required for diphthamide biosynthesis-like 1; DPH1; DPH1 homolog; DPH1 homolog (S. cerevisiae); DPH1-OVCA2; DPH2L; Dph2l1; DPH2-like 1; DPH-like 1; FLJ33211; hypothetical protein LOC550559; mmDph1; ovarian cancer-associated gene 1 protein; Ovarian cancer-associated gene 1 protein homolog; ovarian tumor suppressor candidate 1; Ovca1; RGD1562694; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase 1; Unknown (protein for MGC:127689); zgc:110702 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZG8 | |
DPH1 | |
Recombinant Human OVCA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only