missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Oval PTFE Stir Bar

Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.03960kg

 View more versions of this product

Product Code. 11838952

  • £29.20 / Pack of 10
In Stock
to see stock.
Add to basket

Description

Description

  • Stable, general-purpose magnetic stir bar
  • Provides excellent turbulence for round-bottom flasks and containers
  • Autoclavable
Specifications

Specifications

10 mm
White
Round bottom flasks
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Oval
PTFE encapsulated Alnico V magnet core
5/Pk.
Yes
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers