missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand Stir bar, magnetic, oval, 30mm x 10mm
Brand: Fisherbrand
Stir bar, magnetic, oval, 30mm x 10mm
Code : A0
Additional Details : Weight : 0.06000kg
Brand: Fisherbrand
Stir bar, magnetic, oval, 30mm x 10mm
Code : A0
Additional Details : Weight : 0.06000kg