Learn More
BUCHI Outlet Filter for B-290
Collects residual particles from the cyclone and protects the instrument, user and environment from fine particles. BUCHI™ Outlet Filter for B-290 is recommended for use with all BUCHI B-290 Mini Spray Dryers.
Brand: BUCHI 044754
Code : 62
Additional Details : Weight : 5.18000kg
Description
- Complete Outlet Filter for BUCHI B-290 Mini Spray Dryers
- Glass filter body
- Manometer gauge
- Available with a polyester deep filter and a PTFE filter membrane
Specifications
B-290 Mini Spray Dryers | |
Glass Body, Polyester Deep Filter and a PTFE Filter | |
Polyester deep filter and a PTFE filter membrane | |
Complete Outlet Filter |