missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Otx2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | Otx2 |
---|---|
Clone | 246826 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Description
Otx2 Monoclonal specifically detects Otx2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Otx2 | |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P32243 | |
5015 | |
E. coli-derived recombinant human Otx2 (isoform b) Met1-Leu289 Accession # P32243 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Otx2 in Western blots. |
246826 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
OTX2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only