missing translation for 'onlineSavingsMsg'
Learn More

OSU 6162 hydrochloride, Tocris Bioscience™

Dopamine stabilizer

Brand:  Tocris (Bio-Techne) 259910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461544

  • £151.80 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Dopamine stabilizer; lacks high affinity for various neuroreceptors in vitro (Ki values are 447 and 1305 nM for D2 and D3 receptors respectively and > 1 μM for other targets). Exhibits high D2 receptor occupancy in vivo; highly active on dopamine synthesis and turnover. Induces stabilizing effects on psychomotor function in behavioral tests without inducing hypolocomotion or catalepsy.
Specifications

Chemical Identifiers

156907-84-5
317.872
osu 6162 hydrochloride, osu6162 hydrochloride, 3s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, pnu-96391; pnu-0096391; osu-6162; s---3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride; s,s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, 3s-3-3-methanesulfonylphenyl-1-propylpiperidine hydrochloride, unii-v5sbjbujw6, v5sbjbujw6, osu-6162 hydrochloride, osu6162 hydrochloride hplc, 3-3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride
(3S)-3-(3-methylsulfonylphenyl)-1-propylpiperidine;hydrochloride
C15H24ClNO2S
LEMGVHZVBREXAD-PFEQFJNWSA-N
9836644
CCCN1CCCC(C1)C2=CC(=CC=C2)S(=O)(=O)C.Cl

Specifications

OSU 6162 hydrochloride
156907-84-5
osu 6162 hydrochloride, osu6162 hydrochloride, 3s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, pnu-96391; pnu-0096391; osu-6162; s---3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride; s,s-3-3-methylsulfonyl phenyl-1-propylpiperidine hydrochloride, 3s-3-3-methanesulfonylphenyl-1-propylpiperidine hydrochloride, unii-v5sbjbujw6, v5sbjbujw6, osu-6162 hydrochloride, osu6162 hydrochloride hplc, 3-3-methanesulfonyl-phenyl-1-propyl-piperidine hydrochloride
CCCN1CCCC(C1)C2=CC(=CC=C2)S(=O)(=O)C.Cl
317.872
317.87
10mg
C15H24ClNO2S
LEMGVHZVBREXAD-PFEQFJNWSA-N
(3S)-3-(3-methylsulfonylphenyl)-1-propylpiperidine;hydrochloride
9836644
>98%
Videos