Learn More
OSTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552752
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: REAYKTLSSL YSEMQKMNEL ENKAEPGTHL CIDVEDAMNI TRKLWSRTFN CSVP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 89%.
OSTM1 Polyclonal antibody specifically detects OSTM1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
OSTM1 | |
Polyclonal | |
Unconjugated | |
OSTM1 | |
1200002H13Rik; Chloride channel 7 beta subunit; CLCN7 accessory beta subunit; GAIP interacting protein N terminus; GAIP-interacting protein N terminus; GIPN; GL; grey lethal osteopetrosis; grey-lethal osteopetrosis; Grey-lethal protein; HSPC019; OPTB5; osteopetrosis associated transmembrane protein 1; osteopetrosis associated transmembrane protein 1-like; osteopetrosis-associated transmembrane protein 1; Ostm1; UNQ6098/PRO21201 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28962 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WC4 | |
OSTM1 | |
Recombinant protein corresponding to Human OSTM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only